Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001042565.3 | NP_001036030.1 | 4 | 1688 | |
NM_019653.3 | NP_062627.3 | 9 | 1834 | ||
GenBank mRNA | AK019206.2 | - | 9 | 1812 | |
AK048511.1 | - | 4 | 1688 | ||
AK145974.1 | - | 9 | 1813 | ||
AK146752.1 | - | 9 | 1809 | ||
AK150394.1 | - | 9 | 1809 | ||
AK152545.1 | - | 9 | 1812 | ||
AK159905.1 | - | 9 | 1809 | ||
AK167174.1 | - | 9 | 1811 | ||
AK167394.1 | - | 9 | 1812 | ||
AK167942.1 | - | 9 | 1809 | ||
AK168468.1 | - | 9 | 3232 | ||
AK168565.1 | - | 9 | 3231 | ||
AK169058.1 | - | 9 | 1809 | ||
BC019601.1 | - | 4 | 905 | ||
BC053391.1 | - | 9 | 1585 | 30073116 |