Gene Transcripts

Gene Symbol
Selk
Entrez Gene ID
80795
Gene Name
selenoprotein K
Gene Aliases
1110001C03Rik, AA673253, HSPC030, Hsp30
Chromosome Location
Chr. 14: 29968380 - 29975074 on Build GRCm38
UniGene ID
Mm.104491
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_019979.2 NP_064363.2 5 448
GenBank mRNA AF170920.1 - 5 477
AK003206.1 - 5 376
AK003280.1 - 1 42
AK006674.1 - 5 348
AK007523.1 - 5 360
AK008419.1 - 5 384
BC038049.1 - 5 389 2645414
BC096044.1 - 5 330 1248280
BC096677.1 - 5 378 874920

Gene Symbol
BC052688
Entrez Gene ID
432812
Gene Name
cDNA sequence BC052688
Gene Aliases
Chromosome Location
Chr. 13: 61771580 - 61787282 on Build GRCm38
UniGene ID
Mm.317358
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA BC066226.1 - 2741

Gene Symbol
Gm12060
Entrez Gene ID
100134990
Gene Name
selenoprotein K pseudogene
Gene Aliases
OTTMUSG00000005275
Chromosome Location
Chr. 11: 23558058 - 23558842 on Build GRCm38
UniGene ID
Mm.247328
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA BC027278.1 - 1 422

View Full Product Details