Gene Transcripts

Gene Symbol
MED16
Entrez Gene ID
10025
Gene Name
mediator complex subunit 16
Gene Aliases
DRIP92, THRAP5, TRAP95
Chromosome Location
Chr. 19: 867961 - 893228 on Build GRCh38
UniGene ID
Hs.365207
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_005481.2 NP_005472.2 8 1455
XM_017026120.1 XP_016881609.1 8 1450
XM_017026121.1 XP_016881610.1 7 1295
GenBank mRNA AF106934.1 - 8 1442
AF121228.1 - 7 1304
AL137732.1 - 1 157
BC004554.1 - 8 1436 4053743
BC007853.2 - 4 565
BC011841.1 - 8 1436 4053664
BC017282.2 - 8 1430 4876977
EU392449.1 - 7 1321
EU392451.1 - 6 883
EU392452.1 - 4 604
EU392453.1 - 4 598

View Full Product Details