Gene Transcripts

Gene Symbol
Stard3nl
Entrez Gene ID
76205
Gene Name
STARD3 N-terminal like
Gene Aliases
0610035N01Rik, 6530409L22Rik, AW124774, MENTHO
Chromosome Location
Chr. 13: 19357676 - 19395809 on Build GRCm38
UniGene ID
Mm.41940
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK002760.1 - 7 787
AK152116.1 - 8 850
AK152378.1 - 8 801
BC003334.1 - 8 804 3256725

View Full Product Details