Gene Transcripts

Gene Symbol
Rab22a
Entrez Gene ID
19334
Gene Name
RAB22A, member RAS oncogene family
Gene Aliases
3732413A17Rik, AI662177, AW319644, AW550514, E130120E14Rik, Rab22
Chromosome Location
Chr. 2: 173659845 - 173702182 on Build GRCm38
UniGene ID
Mm.275304
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK014407.1 - 4309
AK031817.1 - 661
AK045756.1 - 407
AK046417.1 - 114
AK147619.1 - 4328

View Full Product Details