Gene Transcripts

Gene Symbol
Mcm8
Entrez Gene ID
66634
Gene Name
minichromosome maintenance deficient 8 (S. cerevisiae)
Gene Aliases
5730432L01Rik
Chromosome Location
Chr. 2: 132816141 - 132844197 on Build GRCm38
UniGene ID
Mm.157070
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001291054.1 NP_001277983.1 6 845
NM_025676.4 NP_079952.2 5 761
GenBank mRNA AK017607.1 - 6 743
AK133858.1 - 6 802
BC046780.1 - 6 792 5716136
BC052070.1 - 5 567 5716125

View Full Product Details