Gene Transcripts

Gene Symbol
Slc16a9
Entrez Gene ID
66859
Gene Name
solute carrier family 16 (monocarboxylic acid transporters), member 9
Gene Aliases
1200003C15Rik, 4930425B13Rik, AI552636
Chromosome Location
Chr. 10: 70245276 - 70285951 on Build GRCm38
UniGene ID
Mm.19325
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_025807.3 NP_080083.2 6 1641
GenBank mRNA AK004556.1 - 6 1642
AK004599.1 - 6 1820
AK004684.2 - 6 1699
AK015196.1 - 4 1418
BC034205.1 - 6 1597 4987055
BC055839.1 - 6 1599 4237811

View Full Product Details