Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001168516.1 | NP_001161988.1 | 3 | 881 | |
NM_001168517.1 | NP_001161989.1 | 3 | 928 | ||
NM_027476.3 | NP_081752.2 | 3 | 1174 | ||
GenBank mRNA | AK017737.1 | - | 3 | 1139 | |
AK028164.1 | - | 3 | 1137 | ||
AK043923.1 | - | 3 | 1131 | ||
AK169337.1 | - | 3 | 1133 | ||
AK170233.1 | - | 3 | 1158 | ||
AK170706.1 | - | 3 | 1158 | ||
AK171568.1 | - | 3 | 1158 | ||
AK172151.1 | - | 3 | 928 | ||
BC056621.1 | - | 3 | 896 | ||
BC071194.1 | - | 3 | 1115 |