Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_144538.2 | NP_653121.1 | 4 | 496 | |
GenBank mRNA | AK004767.1 | - | 4 | 491 | |
AK132764.1 | - | 7 | 1203 | ||
AK161298.1 | - | 7 | 1203 | ||
BC020147.1 | - | 4 | 461 | 3708973 |