Gene Transcripts

Gene Symbol
SH2B3
Entrez Gene ID
10019
Gene Name
SH2B adaptor protein 3
Gene Aliases
IDDM20, LNK
Chromosome Location
Chr. 12: 111405106 - 111451623 on Build GRCh38
UniGene ID
Hs.506784
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001291424.1 NP_001278353.1 7 1178
NM_005475.2 NP_005466.1 8 2101
XM_005253818.4 XP_005253875.1 9 2245
XM_005253819.4 XP_005253876.1 8 2128
XM_006719180.3 XP_006719243.1 7 1730
XM_011537719.2 XP_011536021.1 9 2248
XM_011537720.2 XP_011536022.1 9 1913
XM_011537721.2 XP_011536023.1 7 3713
GenBank mRNA AB208911.1 - 7 1337
AF055581.1 - 8 2101
AK302173.1 - 7 1178
BC136451.1 - 8 1787 9020440

View Full Product Details