Gene Transcripts

Gene Symbol
Cplx4
Entrez Gene ID
225644
Gene Name
complexin 4
Gene Aliases
A930004D23Rik, CPXIV, Gm957
Chromosome Location
Chr. 18: 65955722 - 65970178 on Build GRCm38
UniGene ID
Mm.192285
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_145493.1 NP_663468.1 3 858
GenBank mRNA AK044262.1 - 3 866
BC016082.1 - 3 858 4501240
BC029011.1 - 3 806 5357047

View Full Product Details