Gene Transcripts

Gene Symbol
Casc1
Entrez Gene ID
320662
Gene Name
cancer susceptibility candidate 1
Gene Aliases
A230084G12Rik, Las1
Chromosome Location
Chr. 6: 145174872 - 145210970 on Build GRCm38
UniGene ID
Mm.207779
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_177222.4 NP_796196.3 9 1088
GenBank mRNA AK034148.1 - 8 936
AK132912.1 - 9 1312
AY423542.1 - 9 1019
AY485607.1 - 9 1088
AY485608.1 - 9 1088
BC072568.1 - 7 930
BC120743.1 - 9 992 40129666
BC125423.1 - 9 992 40129938
BC144801.1 - 9 992

View Full Product Details