Gene Transcripts

Gene Symbol
Lrrc4b
Entrez Gene ID
272381
Gene Name
leucine rich repeat containing 4B
Gene Aliases
A830007M12, Lrig4
Chromosome Location
Chr. 7: 44442487 - 44463351 on Build GRCm38
UniGene ID
Mm.44173
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_198250.1 NP_937893.1 3 447
GenBank mRNA AK140418.1 - 2 394
BC053486.1 - 2 311
BC060263.1 - 3 447 5686281

View Full Product Details