Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_201518.4 | NP_958926.1 | 2 | 1480 | |
GenBank mRNA | AK041311.1 | - | 2 | 1453 | |
AK076530.1 | - | 1 | 920 | ||
AK135149.1 | - | 2 | 1481 | ||
AK136973.1 | - | 2 | 1474 | ||
AK140831.1 | - | 2 | 1713 | ||
AK172945.1 | - | 1 | 640 | ||
AY495669.1 | - | 1 | 919 | ||
BC067058.1 | - | 2 | 1461 | ||
BC096471.1 | - | 2 | 1448 | 3582155 | |
BC138297.1 | - | 1 | 928 | 8861316 | |
BC138298.1 | - | 1 | 928 | 8861317 |