Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_010578.2 | NP_034708.1 | 3 | 402 | |
GenBank mRNA | AK088016.1 | - | 3 | 303 | |
AK088729.1 | - | 3 | 149 | ||
AK135521.1 | - | 3 | 301 | ||
AK159689.1 | - | 3 | 244 | ||
AK167682.1 | - | 3 | 300 | ||
BC050906.1 | - | 3 | 275 | 5721348 | |
X15202.1 | - | 2 | 95 | ||
Y00769.1 | - | 3 | 190 |