Gene Transcripts

Gene Symbol
Setd8
Entrez Gene ID
67956
Gene Name
SET domain containing (lysine methyltransferase) 8
Gene Aliases
2410195B05Rik, AA617402, AW536475, PR-SET7
Chromosome Location
Chr. 5: 124439930 - 124462308 on Build GRCm38
UniGene ID
Mm.137966
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_030241.2 NP_084517.2 7, 8 914
GenBank mRNA AK030904.1 - 7, 8 908
BC108333.1 - 7, 8 843 4978041

View Full Product Details