Gene Transcripts

Gene Symbol
Prlhr
Entrez Gene ID
226278
Gene Name
prolactin releasing hormone receptor
Gene Aliases
Gm339, Gpr10, Gr3, Prrpr
Chromosome Location
Chr. 19: 60465641 - 60469431 on Build GRCm38
UniGene ID
Mm.377241
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_201615.2 NP_963909.2 1 1351
GenBank mRNA BC125649.1 - 1 1283
BC125651.1 - 1 1283 40130166

View Full Product Details