Gene Transcripts

Gene Symbol
Vmn1r65
Entrez Gene ID
81013
Gene Name
vomeronasal 1 receptor 65
Gene Aliases
V1rd6, V3R6
Chromosome Location
Chr. 7: 6007750 - 6011010 on Build GRCm38
UniGene ID
Mm.160378
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_030738.2 NP_109663.1 3 1799
GenBank mRNA AF324870.1 - 3 1800

View Full Product Details