Gene Transcripts

Gene Symbol
Cyp2d40
Entrez Gene ID
71754
Gene Name
cytochrome P450, family 2, subfamily d, polypeptide 40
Gene Aliases
1300013D18Rik, CYPIID3, P450-DB3
Chromosome Location
Chr. 15: 82759833 - 82764122 on Build GRCm38
UniGene ID
Mm.443024
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_023623.2 NP_076112.2 3 420
GenBank mRNA AK004984.1 - 3 483
BC061152.1 - 3 420 30308952
BC087906.1 - 3 481 30306972

Gene Symbol
Cyp2d13
Entrez Gene ID
68444
Gene Name
cytochrome P450, family 2, subfamily d, polypeptide 13
Gene Aliases
1300007K12Rik
Chromosome Location
Chr. 15: 82636750 - 82642045 on Build GRCm38
UniGene ID
Mm.443212
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK004933.1 - 796
BC018344.1 - 6 946

View Full Product Details