Gene Transcripts

Gene Symbol
Rhou
Entrez Gene ID
69581
Gene Name
ras homolog gene family, member U
Gene Aliases
2310026M05Rik, AI182090, Arhu, CDC42L1, G28K, WRCH-1, WRCH1, mG28K
Chromosome Location
Chr. 8: 123653929 - 123663880 on Build GRCm38
UniGene ID
Mm.168257
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_133955.4 NP_598716.1 3 2115
GenBank mRNA AB051827.1 - 3 2110
AF378088.1 - 3 2098
AK009513.1 - 1 149

View Full Product Details