Gene Transcripts

Gene Symbol
Tdpoz1
Entrez Gene ID
207213
Gene Name
TD and POZ domain containing 1
Gene Aliases
2cpoz56, Mapp2, Spopl1
Chromosome Location
Chr. 3: 93669333 - 93676283 on Build GRCm38
UniGene ID
Mm.425165
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_148949.2 NP_683751.2 2 1256
GenBank mRNA AF290198.1 - 2 1241
BC125302.1 - 1 1118 40129817
BC125304.1 - 1 1118 40129819

Gene Symbol
Gm9125
Entrez Gene ID
668359
Gene Name
predicted gene 9125
Gene Aliases
EG668359
Chromosome Location
Chr. 3: 94047981 - 94054885 on Build GRCm38
UniGene ID
Mm.80593
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001163730.2 NP_001157202.1 2 1283
GenBank mRNA AK033449.1 - 2 1284

View Full Product Details