Gene Transcripts

Gene Symbol
Alg11
Entrez Gene ID
207958
Gene Name
asparagine-linked glycosylation 11 (alpha-1,2-mannosyltransferase)
Gene Aliases
AI849156, AW492253, B230397C21, Mmat-4(5)
Chromosome Location
Chr. 8: 22060721 - 22071627 on Build GRCm38
UniGene ID
Mm.267439
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001243161.1 NP_001230090.1 3 598
NM_183142.4 NP_898965.1 3 472
GenBank mRNA AB288237.1 - 3 592
AB288238.1 - 3 596
AK046475.1 - 3 474
AK134312.1 - 3 573
AK157746.1 - 3 598
BC061469.1 - 3 581 6821552

View Full Product Details