Gene Transcripts

Gene Symbol
Mdga2
Entrez Gene ID
320772
Gene Name
MAM domain containing glycosylphosphatidylinositol anchor 2
Gene Aliases
6720489L24Rik, 9330209L04Rik, Adp, Mamdc1
Chromosome Location
Chr. 12: 66466059 - 67222549 on Build GRCm38
UniGene ID
Mm.227680
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001193266.1 NP_001180195.2 2 1658
NM_207010.2 NP_996893.3 2 1658
GenBank mRNA AK137667.1 - 2 1659
AY371925.1 - 2 312
BC137901.1 - 2 311 8860920
BC137902.1 - 2 311 8860921
BC145021.1 - 2 311
BC171938.1 - 2 311 9054224

View Full Product Details