Gene Transcripts
- Gene Symbol
- NELFA
- Entrez Gene ID
- 7469
- Gene Name
- negative elongation factor complex member A
- Gene Aliases
- NELF-A, P/OKcl.15, WHSC2
- Chromosome Location
- Chr. 4: 1982714 - 2009972 on Build GRCh38
- UniGene ID
- Hs.21771
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_005663.4 |
NP_005654.3
|
5, 6 |
1034 |
|
XM_017008589.1 |
XP_016864078.1
|
6, 7 |
1855 |
|
GenBank mRNA |
AB044549.1 |
- |
4, 5 |
821 |
|
AF101434.1 |
- |
5, 6 |
763 |
|
AF131751.1 |
- |
1, 2 |
284 |
|
AK001304.1 |
- |
5, 6 |
821 |
|
AK094040.1 |
- |
4, 5 |
594 |
|
AK095737.1 |
- |
4, 5 |
1077 |
|
AK126056.1 |
- |
5, 6 |
1909 |
|
AK222853.1 |
- |
5, 6 |
771 |
|
BC002764.2 |
- |
5, 6 |
774 |
3630594 |
View Full Product Details