Gene Transcripts

Gene Symbol
Cercam
Entrez Gene ID
99151
Gene Name
cerebral endothelial cell adhesion molecule
Gene Aliases
2600006K01Rik, AL024097, Ceecam1
Chromosome Location
Chr. 2: 29845105 - 29882845 on Build GRCm38
UniGene ID
Mm.296336
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_207298.2 NP_997181.1 4 567
GenBank mRNA AK155384.1 - 4 563
BB621603.1 - 4 567
BC052176.1 - 4 528
BC138847.1 - 4 546 8861869

View Full Product Details