Gene Transcripts
- Gene Symbol
- PTPRCAP
- Entrez Gene ID
- 5790
- Gene Name
- protein tyrosine phosphatase, receptor type C associated protein
- Gene Aliases
- CD45-AP, LPAP
- Chromosome Location
- Chr. 11: 67435510 - 67437682 on Build GRCh38
- UniGene ID
- Hs.155975
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_005608.2 |
NP_005599.1
|
2 |
881 |
|
GenBank mRNA |
X81422.1 |
- |
2 |
880 |
|
View Full Product Details