Gene Transcripts
- Gene Symbol
- RPS14
- Entrez Gene ID
- 6208
- Gene Name
- ribosomal protein S14
- Gene Aliases
- EMTB, S14
- Chromosome Location
- Chr. 5: 150444229 - 150449810 on Build GRCh38
- UniGene ID
- Hs.744846
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001025070.1 |
NP_001020241.1
|
3, 4 |
576 |
|
NM_001025071.1 |
NP_001020242.1
|
3, 4 |
582 |
|
NM_005617.3 |
NP_005608.1
|
3, 4 |
365 |
|
XM_017009684.1 |
XP_016865173.1
|
3, 4 |
419 |
|
GenBank mRNA |
AA467888.1 |
- |
1, 2 |
132 |
|
AK312179.1 |
- |
3, 4 |
344 |
|
BC001126.1 |
- |
1 |
328 |
2967098 |
BC003401.1 |
- |
2, 3 |
322 |
3448752 |
BC006784.1 |
- |
1 |
333 |
2900907 |
BC020515.1 |
- |
3, 4 |
515 |
3856643 |
BC091474.1 |
- |
3, 4 |
346 |
30334486 |
BM454896.1 |
- |
3, 4 |
545 |
|
BM557014.1 |
- |
3, 4 |
517 |
|
- Gene Symbol
- RPS14P3
- Entrez Gene ID
- 644068
- Gene Name
- ribosomal protein S14 pseudogene 3
- Gene Aliases
- RPS14_1_16
- Chromosome Location
- Chr. 1: 19607807 - 19608644 on Build GRCh38
- UniGene ID
- Hs.660760
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_077246.1 |
|
1 |
616 |
|
GenBank mRNA |
BC072682.1 |
- |
1 |
616 |
|
BU176905.1 |
- |
1 |
584 |
|
View Full Product Details