Gene Transcripts
- Gene Symbol
- Lrrd1
- Entrez Gene ID
- 242838
- Gene Name
- leucine rich repeats and death domain containing 1
- Gene Aliases
- 4932412H11Rik
- Chromosome Location
- Chr. 5: 3845173 - 3866596 on Build GRCm38
- UniGene ID
- Mm.482273
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK029981.1 |
- |
2 |
698 |
|
View Full Product Details