Gene Transcripts

Gene Symbol
Fam71e2
Entrez Gene ID
243822
Gene Name
family with sequence similarity 71, member E2
Gene Aliases
4930401F20Rik
Chromosome Location
Chr. 7: 4753226 - 4771361 on Build GRCm38
UniGene ID
Mm.333542
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_172895.3 NP_766483.3 3 675
GenBank mRNA AK076650.1 - 3 733

View Full Product Details