Gene Transcripts

Gene Symbol
Ssc5d
Entrez Gene ID
269855
Gene Name
scavenger receptor cysteine rich family, 5 domains
Gene Aliases
A430110N23Rik, S5D-SRCRB
Chromosome Location
Chr. 7: 4925130 - 4944828 on Build GRCm38
UniGene ID
Mm.44190
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_173008.2 NP_766596.1 14 3405
GenBank mRNA AK079906.1 - 14 3406
AK165666.1 - 14 3467
BC141044.1 - 14 3403 9055656
EU850434.1 - 14 3406

View Full Product Details