Gene Transcripts

Gene Symbol
Sspo
Entrez Gene ID
243369
Gene Name
SCO-spondin
Gene Aliases
C79529, Scospondin
Chromosome Location
Chr. 6: 48446243 - 48501262 on Build GRCm38
UniGene ID
Mm.25039
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_173428.3 NP_775604.3 2 136
GenBank mRNA AJ491857.1 - 2 136

View Full Product Details