Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001290377.1 | NP_001277306.1 | 4 | 660 | |
NM_175632.3 | NP_783440.1 | 4 | 678 | ||
GenBank mRNA | AF391159.1 | - | 4 | 636 | |
AF391160.1 | - | 4 | 660 | ||
AF391161.1 | - | 4 | 654 | ||
BC137776.1 | - | 4 | 605 | 8860795 |