Gene Transcripts

Gene Symbol
Mical2
Entrez Gene ID
320878
Gene Name
microtubule associated monooxygenase, calponin and LIM domain containing 2
Gene Aliases
5330438E18Rik, 9530064J02, MICAL-2, mKIAA0750
Chromosome Location
Chr. 7: 112225836 - 112355194 on Build GRCm38
UniGene ID
Mm.488195
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001193305.1 NP_001180234.1 14 2074
NM_177282.5 NP_796256.1 14 2074
GenBank mRNA AK030608.1 - 14 2055
AK053370.1 - 6 3148
AK138935.1 - 14 2048
AK143191.1 - 14 2021
AK161075.1 - 14 2021
AK163078.1 - 14 2041
AK220353.1 - 12 1751
BC111895.1 - 12 1798 40061129
BC112415.1 - 12 1798 40061128

View Full Product Details