Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_177307.3 | NP_796281.1 | 14 | 2015 | |
GenBank mRNA | AK028950.1 | - | 14 | 2016 | |
AK037192.1 | - | 14 | 2016 | ||
BC052147.1 | - | 1 | 560 | ||
BC145755.1 | - | 14 | 2012 | 40131058 |