Gene Transcripts
- Gene Symbol
- Slc2a12
- Entrez Gene ID
- 353169
- Gene Name
- solute carrier family 2 (facilitated glucose transporter), member 12
- Gene Aliases
- GLUT-12, Glut12
- Chromosome Location
- Chr. 10: 22612723 - 22703880 on Build GRCm38
- UniGene ID
- Mm.473705
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_178934.4 |
NP_849265.2
|
1, 2 |
311 |
|
GenBank mRNA |
AJ549317.2 |
- |
1, 2 |
311 |
|
AK028970.1 |
- |
1, 2 |
284 |
|
AK031659.1 |
- |
1, 2 |
229 |
|
AK035972.1 |
- |
1, 2 |
192 |
|
AK143263.1 |
- |
1, 2 |
300 |
|
AY230881.1 |
- |
1, 2 |
210 |
|
BC096454.1 |
- |
1, 2 |
242 |
4223231 |
BC116314.1 |
- |
1, 2 |
172 |
40095492 |
BC116315.1 |
- |
1, 2 |
175 |
40095493 |
BC138316.1 |
- |
1, 2 |
158 |
8861336 |
BC138317.1 |
- |
1, 2 |
158 |
8861337 |
View Full Product Details