Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001167743.1 | NP_001161215.1 | 4 | 2800 | |
NM_181545.4 | NP_853523.2 | 6 | 3728 | ||
GenBank mRNA | AK150875.1 | - | 4 | 3050 | |
AY261798.1 | - | 6 | 3728 | ||
AY261799.1 | - | 6 | 3690 | ||
BC024709.1 | - | 2 | 1558 |