Gene Transcripts

Gene Symbol
Syt14
Entrez Gene ID
329324
Gene Name
synaptotagmin XIV
Gene Aliases
B230320I09Rik, sytXIV
Chromosome Location
Chr. 1: 192891233 - 193035862 on Build GRCm38
UniGene ID
Mm.170401
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001301370.1 NP_001288299.1 3 338
NM_181546.3 NP_853524.1 3 338
GenBank mRNA AB102947.1 - 3 261
AK045901.1 - 3 339
BC141262.1 - 3 280 9055874
BC145602.1 - 3 280 9054168

View Full Product Details