Gene Transcripts

Gene Symbol
Foxo6
Entrez Gene ID
329934
Gene Name
forkhead box O6
Gene Aliases
Chromosome Location
Chr. 4: 120267078 - 120287261 on Build GRCm38
UniGene ID
Mm.277090
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_194060.1 NP_918949.1 2 1604
GenBank mRNA AJ571690.1 - 2 1604

View Full Product Details