Gene Transcripts

Gene Symbol
Dbpht2
Entrez Gene ID
386753
Gene Name
DNA binding protein with his-thr domain
Gene Aliases
Spot2
Chromosome Location
Chr. 12: 74297474 - 74300468 on Build GRCm38
UniGene ID
Mm.341686
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_198866.2 NP_942566.1 1 1849
GenBank mRNA AF547221.1 - 1 864
AK158661.1 - 1 258

View Full Product Details