Gene Transcripts

Gene Symbol
Znrf2
Entrez Gene ID
387524
Gene Name
zinc and ring finger 2
Gene Aliases
1190002C14Rik, AU044205, D6Ertd365e
Chromosome Location
Chr. 6: 54816916 - 54890224 on Build GRCm38
UniGene ID
Mm.286149
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_199143.2 NP_954594.1 5 999
GenBank mRNA AF513708.1 - 5 999

View Full Product Details