Gene Transcripts

Gene Symbol
Ttc7
Entrez Gene ID
225049
Gene Name
tetratricopeptide repeat domain 7
Gene Aliases
1110035E02Rik, 1700007L07Rik, Ttc7a, fsn, hea
Chromosome Location
Chr. 17: 87282082 - 87381770 on Build GRCm38
UniGene ID
Mm.77396
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_028639.3 NP_082915.1 2 586
GenBank mRNA AK040578.1 - 2 589
AK040610.1 - 2 556
AK050263.1 - 2 573
AK080284.1 - 2 565
AK153704.1 - 2 568
BC042512.1 - 2 263 4223801

View Full Product Details