Gene Transcripts

Gene Symbol
Ncaph
Entrez Gene ID
215387
Gene Name
non-SMC condensin I complex, subunit H
Gene Aliases
A730011O11Rik, Brrn1, HCAP-H, mKIAA0074
Chromosome Location
Chr. 2: 127103809 - 127133954 on Build GRCm38
UniGene ID
Mm.29786
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_144818.3 NP_659067.2 8, 9 1063
GenBank mRNA AK028919.1 - 8, 9 1048
AK042706.1 - 8, 9 980
AK080433.1 - 8, 9 1052
AK129047.1 - 5, 6 616
BC021499.1 - 8, 9 1037 5346298
BC025060.1 - 8, 9 988 3673890

View Full Product Details