Gene Transcripts

Gene Symbol
Klhl20
Entrez Gene ID
226541
Gene Name
kelch-like 20
Gene Aliases
AI504637, D930050H05Rik, Kleip, mKIAA4210
Chromosome Location
Chr. 1: 161088375 - 161131508 on Build GRCm38
UniGene ID
Mm.255165
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001039482.1 NP_001034571.1 3 284
GenBank mRNA AK053102.1 - 3 234
AK053371.1 - 3 248
AK054169.1 - 3 252
AK220547.1 - 3 263
BC019571.1 - 3 256 4235202
BC060183.1 - 3 213

View Full Product Details