Gene Transcripts

Gene Symbol
Blvrb
Entrez Gene ID
233016
Gene Name
biliverdin reductase B (flavin reductase (NADPH))
Gene Aliases
Chromosome Location
Chr. 7: 27447978 - 27465981 on Build GRCm38
UniGene ID
Mm.24021
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001290525.1 NP_001277454.1 5 631
NM_144923.3 NP_659172.1 5 717
GenBank mRNA AK153153.1 - 5 638
AK164759.1 - 1 56
BC006617.1 - 5 657 3967600
BC027279.1 - 5 717 3492861

View Full Product Details