Gene Transcripts

Gene Symbol
Pex6
Entrez Gene ID
224824
Gene Name
peroxisomal biogenesis factor 6
Gene Aliases
AI132582, D130055I09Rik, mKIAA4177
Chromosome Location
Chr. 17: 46711463 - 46725541 on Build GRCm38
UniGene ID
Mm.299399
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_145488.1 NP_663463.1 4, 5 1268
GenBank mRNA AK143173.1 - 4, 5 1256
AK220525.1 - 4, 5 1249
BC003424.1 - 4, 5 1268 2609884

View Full Product Details