Gene Transcripts
- Gene Symbol
- Ina
- Entrez Gene ID
- 226180
- Gene Name
- internexin neuronal intermediate filament protein, alpha
- Gene Aliases
- AV028420, NF-66, NF66
- Chromosome Location
- Chr. 19: 47014698 - 47024358 on Build GRCm38
- UniGene ID
- Mm.276251
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
L36390.1 |
- |
3 |
1265 |
|
View Full Product Details