Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001110212.2 | NP_001103682.1 | 26 | 3537 | |
NM_146112.4 | NP_666224.3 | 26 | 3555 | ||
GenBank mRNA | AK122337.1 | - | 17 | 2626 | |
AY176043.1 | - | 26 | 3501 | ||
BC027137.1 | - | 6 | 900 | ||
BC076610.1 | - | 26 | 3500 | ||
BC082811.1 | - | 7 | 1011 | ||
BC089036.1 | - | 26 | 3511 | 30695300 |