Gene Transcripts

Gene Symbol
Ddx27
Entrez Gene ID
228889
Gene Name
DEAD (Asp-Glu-Ala-Asp) box polypeptide 27
Gene Aliases
C86129
Chromosome Location
Chr. 2: 167015255 - 167034950 on Build GRCm38
UniGene ID
Mm.295031
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA BC024730.1 - 8 1757
BC024981.1 - 8 1757
BC033398.1 - 8 1757

View Full Product Details