Gene Transcripts

Gene Symbol
Kctd11
Entrez Gene ID
216858
Gene Name
potassium channel tetramerisation domain containing 11
Gene Aliases
AF465352, Ren
Chromosome Location
Chr. 11: 69878264 - 69880985 on Build GRCm38
UniGene ID
Mm.239498
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_153143.4 NP_694783.1 1 1553
GenBank mRNA AF465352.1 - 1 1520
AK039558.1 - 1975
AK161602.1 - 1 1548
AK170258.1 - 1 1447
BC052502.1 - 1 801 5119264
BC052734.1 - 1 1504 6816736
BC063052.1 - 1 1504

View Full Product Details