Gene Transcripts

Gene Symbol
AKAP8
Entrez Gene ID
10270
Gene Name
A-kinase anchoring protein 8
Gene Aliases
AKAP 95, AKAP-8, AKAP-95, AKAP95
Chromosome Location
Chr. 19: 15353521 - 15379815 on Build GRCh38
UniGene ID
Hs.594496
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_005858.3 NP_005849.1 10, 11 1380
XM_011527624.2 XP_011525926.1 10, 11 1473
XM_017026141.1 XP_016881630.1 10, 11 1525
GenBank mRNA AK303640.1 - 7, 8 1007
AK308251.1 - 9, 10 1274
AK309529.1 - 7, 8 908
AL050160.1 - 4, 5 308
BC037270.2 - 10, 11 1370 4837309
Y11997.1 - 10, 11 1371

View Full Product Details